565504
Salicylideneaniline
97%
Synonym(s):
2-Hydroxybenzylideneaniline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
mp
49-53 °C (lit.)
SMILES string
Oc1ccccc1\C=N\c2ccccc2
InChI
1S/C13H11NO/c15-13-9-5-4-6-11(13)10-14-12-7-2-1-3-8-12/h1-10,15H/b14-10+
InChI key
QIYHCQVVYSSDTI-GXDHUFHOSA-N
Gene Information
human ... CYP2D6(1565)
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Molecules (Basel, Switzerland), 24(4) (2019-03-01)
Salicylideneaniline (SA) sorbed in cation-exchanged M-ZSM-5 (M = H⁺, Li⁺, Na⁺, K⁺, Rb⁺, Cs⁺ and Zn2+) zeolites was studied by spectroscopic techniques assisted by quantum-chemical calculations. The nature of extra-framework cations present in the zeolite void was found to affect
Journal of fluorescence, 29(1), 111-120 (2018-10-29)
Five luminescent Eu(III)-4-fluoro-N-salicylideneaniline (Fsa) complexes with different N,N'- donor heterocyclic ligands (L) were synthesized (where L = 2,2'-bipyridine (bipy), 4,4'-dimethoxy-2,2'-bipyridine (dmbp), 1,10-phenanthroline (phen), 2,9-dimethyl-1,10-phenanthroline (neo) and 2,2'-biquinoline (biq)) and characterized by structural, thermal and spectroscopic analyses. The photophysical studies of the complexes
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service