Skip to Content
Merck
All Photos(2)

Key Documents

563080

Sigma-Aldrich

Methyl 2-aminothiophene-3-carboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H2SCOOCH3NH2
CAS Number:
Molecular Weight:
157.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

76-81 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1ccsc1N

InChI

1S/C6H7NO2S/c1-9-6(8)4-2-3-10-5(4)7/h2-3H,7H2,1H3

InChI key

DGGJQLCAYQCPDD-UHFFFAOYSA-N

Application

Methyl 2-aminothiophene-3-carboxylate may be used to prepare 3-thiaisatoic anhydride via hydrolysis, followed by the reaction with phosgene.[1]
Methyl 2-aminothiophene-3-carboxylate may be used to synthesize the following:
  • thieno[2,3-d]pyrimidine-2,4(1H,3H)-dione[2][3]
  • thieno[2,3-d]pyrimidin-4(3H)one via reaction with formamide[4]
  • methyl 2-(2,5-dimethyl-1H-pyrrol-1-yl)thiophene-3-carboxylate via reaction with hexane-2,5-dione[5]


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Design, synthesis and biological evaluation of 4-anilinothieno [2, 3-d] pyrimidine-based hydroxamic acid derivatives as novel histone deacetylase inhibitors"
Zhu W, et al.
Bioorganic & Medicinal Chemistry, 22(21), 6146-6155 (2014)
"Chemistry of Ring-Fused Oxazine-2, 4-diones"
lette Y, et al. et al.
European Journal of Organic Chemistry, 2009(21), 3487-3503 (2009)
"The highly potent and selective dipeptidyl peptidase IV inhibitors bearing a thienopyrimidine scaffold effectively treat type 2 diabetes"
Deng J, et al.
European Journal of Medicinal Chemistry, 46(1), 71- 76 (2011)
?Synthesis, immunosuppressive activity and structure?activity relationship study of a new series of 4-N-piperazinyl-thieno [2, 3-d] pyrimidine analogues"
Jang Y-M, et al.
Bioorganic & Medicinal Chemistry Letters, 20(3), 844-847 (2010)
"Synthesis of novel 5-chlorinated 2-aminothiophenes using 2, 5-dimethylpyrrole as an amine protecting group"
Puterova Z, et al.
Journal of Heterocyclic Chemistry, 45(1), 201-207 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service