Skip to Content
Merck
All Photos(1)

Key Documents

554510

Sigma-Aldrich

(R)-(+)-1-Benzyl-2,2-diphenylethylamine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
CHF 189.00

CHF 189.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
CHF 189.00

About This Item

Linear Formula:
(C6H5)2CHCH(NH2)CH2C6H5
CAS Number:
Molecular Weight:
287.40
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

CHF 189.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

optical activity

[α]20/D +8.0°, c = 1% in chloroform

mp

74-78 °C (lit.)

functional group

amine
phenyl

SMILES string

N[C@H](Cc1ccccc1)C(c2ccccc2)c3ccccc3

InChI

1S/C21H21N/c22-20(16-17-10-4-1-5-11-17)21(18-12-6-2-7-13-18)19-14-8-3-9-15-19/h1-15,20-21H,16,22H2/t20-/m1/s1

InChI key

HPRWGZYKYRRJNU-HXUWFJFHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service