Skip to Content
Merck
All Photos(1)

Documents

551007

Sigma-Aldrich

2,7-Di-tert-butyl-9-fluorenylmethyl chloroformate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOOCH2CH(C6H3C(CH3)3)2
CAS Number:
Molecular Weight:
370.91
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

66-70 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)c1ccc-2c(c1)C(COC(Cl)=O)c3cc(ccc-23)C(C)(C)C

InChI

1S/C23H27ClO2/c1-22(2,3)14-7-9-16-17-10-8-15(23(4,5)6)12-19(17)20(18(16)11-14)13-26-21(24)25/h7-12,20H,13H2,1-6H3

InChI key

PHUMWWRUIGKGNN-UHFFFAOYSA-N

General description

2,7-Di-tert-butyl-9-fluorenylmethyl chloroformate contains 2,7-di-tert-butyl-Fmoc protecting group (Fmoc= fluorenylmethoxycarbonyl), which is more soluble in organic solvents when compared to its Fmoc analog. It can be prepared from fluorene in three steps, by di-tert-butylation, hydroxymethylation and phosgenation.

Application

2,7-Di-tert-butyl-9-fluorenylmethyl chloroformate may be used in the preparation of polymer-supported (2,7-di-tert-butyl-9-fluorenyl)methyl succinimidyl carbonate (Dtb-Fmoc-P-OSu), a polymeric reagent for protecting amino groups.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

2,7-Di-tert-butyl-Fmoc-P-OSu: A new polymer-Supported reagent for the protection of the amino group.
Chinchilla R, et al.
Bioorganic & Medicinal Chemistry Letters, 12(14), 1817-1820 (2002)
Fmoc: a more soluble analogue of the 9-fluorenylmethoxycarbonyl protecting group.
K D Stigers et al.
The Journal of organic chemistry, 65(12), 3858-3860 (2000-06-24)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service