Skip to Content
Merck
All Photos(1)

Documents

548464

Sigma-Aldrich

Copper(II) telluride

99.9% trace metals basis

Synonym(s):

Copper monotelluride, Cupric telluride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuTe
CAS Number:
Molecular Weight:
191.15
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.9% trace metals basis

form

crystalline

density

7.100 g/mL at 25 °C (lit.)

SMILES string

[Cu]=[Te]

InChI

1S/Cu.Te

InChI key

QZCHKAUWIRYEGK-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service