Skip to Content
Merck
All Photos(1)

Key Documents

542733

Sigma-Aldrich

4-Fluorobenzenesulfonamide

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4SO2NH2
CAS Number:
Molecular Weight:
175.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

124-127 °C (lit.)

SMILES string

NS(=O)(=O)c1ccc(F)cc1

InChI

1S/C6H6FNO2S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,(H2,8,9,10)

InChI key

LFLSATHZMYYIAQ-UHFFFAOYSA-N

General description

4-Fluorobenzenesulfonamide is a para-halogen benzenesulfonamide. Ab initio Hartree-Fock (HF) and density functional theory (DFT) have been used to investigate the structural features of 4-fluorobenzenesulfonamide.[1]

Application

4-Fluorobenzenesulfonamide may be used to synthesize 4-fluorophenylsulfonyldithiocarbimate potassium dihydrate[2] and amino-substituted sulfanilamide derivatives.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guillaume Compain et al.
Bioorganic & medicinal chemistry, 21(6), 1555-1563 (2012-06-19)
A series of new, halogen containing N-substituted 4-aminobenzenesulfonamides were synthesized by using superacid HF/SbF5 chemistry and investigated as inhibitors of several human carbonic anhydrase (hCA, EC 4.2.1.1) isoforms, that is, the cytosolic hCA I and II and, the tumor-associated transmembrane
L B Dugad et al.
Biochemistry, 27(12), 4310-4316 (1988-06-14)
Binding of 4-fluorobenzenesulfonamide to human carbonic anhydrases I and II has been studied by proton, fluorine, and nitrogen-15 nuclear magnetic resonance spectroscopy. All three types of experiments provide evidence that the stoichiometry of the interaction of this inhibitor with both
Bis (4-fluorophenylsulfonyldithiocarbimato) zincate (ii) salts: new antifungals for the control of botrytis blight.
Oliveira AA, et al.
Quimica nova, 38(6), 757-761 (2015)
Theoretical investigation on the molecular structure, Infrared, Raman and NMR spectra of para-halogen benzenesulfonamides, 4-XC6H4SO2NH2 (X= Cl, Br or F).
Karabacak M, et al.
Journal of Molecular Structure, 919(1), 26-33 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service