Skip to Content
Merck
All Photos(2)

Key Documents

541524

Sigma-Aldrich

5,5-Dimethyl-1,3,2-dioxaphosphorinan-2-one

96%

Synonym(s):

2,2-Dimethylpropane-1,3-diol cyclic phosphonate, Cyclic 2,2-dimethyltrimethylene phosphonate, Neopentylene phosphite, Neopentylglycol hydrogen phosphite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11O3P
CAS Number:
Molecular Weight:
150.11
EC Number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

51-56 °C (lit.)

SMILES string

CC1(C)CO[PH](=O)OC1

InChI

1S/C5H11O3P/c1-5(2)3-7-9(6)8-4-5/h9H,3-4H2,1-2H3

InChI key

QHXZLTUNAVKFIT-UHFFFAOYSA-N

Application

Reactant for:
  • Palladium(I) catalyzed addition of P(O)-H bonds
  • Highly regioselective modified Pudovik addition reaction
  • Radical reactions mediated by manganese(III) acetate
  • Stable cyclic DEPMPO derivatives as mechanistic markers of stereoselective hydroxyl radical adduct formation in biological systems
  • Copper-catalyzed aerobic oxidative dehydrogenative coupling reactions
  • Bisphosphonates/phosphanamidates as anticancer agents

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.9 °F - closed cup

Flash Point(C)

109.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Fukal et al.
Physical chemistry chemical physics : PCCP, 21(19), 9924-9934 (2019-05-01)
Structural interpretation of the 31P NMR shifts measured in O,O-diethyl thiophosphate (PT), 5,5-dimethyl-2-mercapto-1,3,2-dioxaphosphorinane 2-oxide (cPT), diethylphosphate (P) and 5,5-dimethyl-2-hydroxy-1,3,2-dioxaphosphinane 2-oxide (cP) was obtained by means of theoretical calculations including the effects of geometry, molecular dynamics, and solvent, relativistic effects and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service