Skip to Content
Merck
All Photos(1)

Key Documents

540714

Sigma-Aldrich

Tetrahydrothiopyran-4-ol

97%

Synonym(s):

4-Tetrahydrothiopyranol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10OS
CAS Number:
Molecular Weight:
118.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

47-51 °C (lit.)

SMILES string

OC1CCSCC1

InChI

1S/C5H10OS/c6-5-1-3-7-4-2-5/h5-6H,1-4H2

InChI key

YODQQARABJQLIP-UHFFFAOYSA-N

General description

Tetrahydrothiopyran-4-ol is a sulfur containing heterocyclic building block. Tetrahydrothiopyran-4-ol can be synthesized from tetrahydrothiopyran-4-one.[1] It participates in the synthesis of 4-hydroxyl-1-methyltetrahydrothiopyranium iodide.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potential antihistamine agents: 4-(6, 11-Dihydrodibenzo [b, e] thiepin-11-ylidene)-1-methyltetrahydrothiopyranium iodide and similar sulfonium salts derived from related tricyclic systems.
Polivka Z, et al.
Collection of Czechoslovak Chemical Communications, 52(11), 2758-2774 (1987)
Kagakkai N.
Bulletin of the Chemical Society of Japan, 68, 2747-2747 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service