Skip to Content
Merck
All Photos(2)

Documents

538906

Sigma-Aldrich

5-Amino-2-pyridinecarbonitrile

96%

Synonym(s):

5-Amino-2-cyanopyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N3
CAS Number:
Molecular Weight:
119.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

148-152 °C (lit.)

SMILES string

Nc1ccc(nc1)C#N

InChI

1S/C6H5N3/c7-3-6-2-1-5(8)4-9-6/h1-2,4H,8H2

InChI key

IFOXWHQFTSCNQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Amino-2-pyridinecarbonitrile (5-Amino-2-cyanopyridine) may be used to synthesize 5-amino-2-pyridine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of dye structure on the dyeing and optical properties of dichroic dyes for PVA polarizing film
Chang JB, et al.
Dyes and Pigments, 88.3, 366-371 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service