Skip to Content
Merck
All Photos(2)

Key Documents

533645

Sigma-Aldrich

2,2′-Difluorobiphenyl

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 375.00

CHF 375.00


Estimated to ship on05 April 2025


Request a Bulk Order

Select a Size

Change View
5 G
CHF 375.00

About This Item

Linear Formula:
FC6H4C6H4F
CAS Number:
Molecular Weight:
190.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 375.00


Estimated to ship on05 April 2025


Request a Bulk Order

Quality Level

Assay

98%

mp

117-121 °C (lit.)

functional group

fluoro

SMILES string

Fc1ccccc1-c2ccccc2F

InChI

1S/C12H8F2/c13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14/h1-8H

InChI key

PXFIPIAXFGAEMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service