Skip to Content
Merck
All Photos(1)

Key Documents

532592

Sigma-Aldrich

4-Methyl-3-nitropyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6N2O2
CAS Number:
Molecular Weight:
138.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

refractive index

n20/D 1.549

bp

238 °C

mp

24-28 °C

density

1.228 g/mL at 25 °C

SMILES string

Cc1ccncc1[N+]([O-])=O

InChI

1S/C6H6N2O2/c1-5-2-3-7-4-6(5)8(9)10/h2-4H,1H3

InChI key

JLNRJMGYBKMDGI-UHFFFAOYSA-N

General description

4-Methyl-3-nitropyridine, a substituted nitropyridine, can be prepared by the malonation of 4-methoxy-3-nitropyridine with sodium diethyl malonate.[1]

Application

4-Methyl-3-nitropyridine may be used as a starting material in the synthesis of ethyl 6-azaindole-2-carboxylate and also 3-substituted azaindoles.[2]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

226.9 °F - closed cup

Flash Point(C)

108.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and reactivity of 4-, 5-and 6-azaindoles.
Popowycz F, et al.
Tetrahedron, 63(36), 8689-8707 (2007)
Pyridine derivatives: PART VI Malonations of substituted nitropyridines.
Gruber W.
Canadian Journal of Chemistry, 31(12), 1181-1188 (1953)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service