Skip to Content
Merck
All Photos(1)

Key Documents

532223

Sigma-Aldrich

5-Amino-1,3-dimethylpyrazole

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3
CAS Number:
Molecular Weight:
111.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

65-69 °C (lit.)

SMILES string

Cc1cc(N)n(C)n1

InChI

1S/C5H9N3/c1-4-3-5(6)8(2)7-4/h3H,6H2,1-2H3

InChI key

ZFDGMMZLXSFNFU-UHFFFAOYSA-N

General description

5-Amino-1,3-dimethylpyrazole undergoes cyclocondensation with ethyl acetoacetate to form the corresponding tetrahydropyrazolopyridine derivatives.

Application

5-Amino-1,3-dimethylpyrazole may be used in the preparation of:
  • 5-benzamido-1,3-dimethylpyrazole
  • diethyl 2-{[(1,3-dimethyl-1H-pyrazol-5-yl)amino]methylene}malonate
  • 5-amino-1,3-dimethyl-4-phthalidylpyrazole
  • (E)-N-(3,7-dimethylocta-2,6-dienyl)-1,3-dimethyl-1H-pyrazol-5-amine analog(LQFM002)
  • 4-isopropyl-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridin-6-ol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The reaction of o?phthalaldchydic acid with 5?amino?1,3?dimethylpyrazole.
Swett LR and Aynilian GH.
Journal of Heterocyclic Chemistry, 12(6), 1135-1136 (1975)
1,3-Oxazines and related compounds. II.1) Ring contraction reaction of 1, 3-oxazin-4-one derivatives into 1,2,4-triazoles and pyrazoles.
Yamamoto, et al.
Chemical & Pharmaceutical Bulletin, 26(6), 1825-1831 (1978)
E A Costa et al.
Life sciences, 92(3), 237-244 (2013-01-09)
The current study describes the synthesis and pharmacological evaluation of (E)-N-(3,7-dimethylocta-2,6-dienyl)-1,3-dimethyl-1H-pyrazol-5-amine (LQFM002), a compound originally designed through a molecular simplification strategy from 4-nerolidylcatechol. LQFM002 was evaluated for preservation of the PLA(2) enzyme inhibitory effects of the lead compound, 4-nerolidylcatechol, using
The cyclocondensation of 5?amino?1,3?dimethylpyrazole with ethyl acetoacetate. Synthesis of isomeric pyrazolopyridones.
Ratajczyk JD and Swett LR.
Journal of Heterocyclic Chemistry, 12(3), 517-522 (1975)
Hiroshi Ochiai et al.
Chemical & pharmaceutical bulletin, 52(9), 1098-1104 (2004-09-02)
A series of 4-anilinopyrazolopyridine derivatives were synthesized and biologically evaluated as inhibitors of phosphodiesterase (PDE4). Chemical modification of 3, a structurally new chemical lead that was found in our in-house library, was focused on 1- and 3-substituents. Full details of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service