Skip to Content
Merck
All Photos(1)

Key Documents

531359

Sigma-Aldrich

Fmoc-Glycine, polymer-bound on Wang resin

extent of labeling: 1.0 mmol/g loading, 1 % cross-linked

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H29NO5
CAS Number:
Molecular Weight:
507.58
UNSPSC Code:
12352200
PubChem Substance ID:

crosslinking

1 % cross-linked

extent of labeling

1.0 mmol/g loading

application(s)

peptide synthesis

functional group

Fmoc

SMILES string

P=S.Cc1ccc(COc2ccc(COC(=O)CNC(=O)OCC3c4ccccc4-c5ccccc35)cc2)cc1

InChI

1S/C17H15NO4/c19-16(20)9-18-17(21)22-10-15-13-7-3-1-5-11(13)12-6-2-4-8-14(12)15/h1-8,15H,9-10H2,(H,18,21)(H,19,20)

InChI key

NDKDFTQNXLHCGO-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K R Sunil Kumar et al.
ACS nano, 8(5), 4157-4165 (2014-04-01)
To apply motor proteins as natural nanomolecular machines to transporting systems in nanotechnology, complete temporal control over ON/OFF switching of the motility is necessary. We have studied the photoresponsive inhibition properties of azobenzene-tethered peptides for regulation of kinesin-microtubule motility. Although
Yun Song et al.
Cell reports, 30(8), 2699-2711 (2020-02-27)
The transcriptional corepressor complex CoREST is one of seven histone deacetylase complexes that regulate the genome through controlling chromatin acetylation. The CoREST complex is unique in containing both histone demethylase and deacetylase enzymes, LSD1 and HDAC1, held together by the
Poonam Saraf et al.
Pharmaceutical research, 32(9), 3087-3101 (2015-06-13)
The objective of this work was to compare the efficacy of self-assembling cyclic and linear RGD peptide amphiphiles as carriers for delivering paclitaxel to αvβ3 integrin overexpressing tumors. Linear (C18-ADA5-RGD) and cyclic (C18-ADA5-cRGDfK) peptide amphiphiles were synthesized and characterized for
Sachiro Kakinoki et al.
Acta biomaterialia, 13, 42-51 (2014-12-03)
Mechanotransduction in the regulation of cellular responses has been previously studied using elastic hydrogels. Because cells interact only with the surface of biomaterials, we are focusing on the molecular mobility at the outermost surface of biomaterials. In this study, surfaces
Yang Li et al.
Macromolecular bioscience, 15(1), 52-62 (2014-12-06)
To address the downside of conventional photo-patterning which can alter the chemical composition of protein scaffolds, we developed a non-covalent photo-patterning strategy for gelatin (denatured collagen) hydrogels that utilizes UV activated triple helical hybridization of caged collagen mimetic peptide (caged

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service