Skip to Content
Merck
All Photos(2)

Key Documents

524042

Sigma-Aldrich

4-(tert-Butyldimethylsilyloxy)phenylboronic acid

≥95%

Synonym(s):

4-(tert-Butyldimethylsilyloxy)benzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2OC6H4B(OH)2
CAS Number:
Molecular Weight:
252.19
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

≥95%

form

solid

mp

194-198 °C (lit.)

SMILES string

CC(C)(C)[Si](C)(C)Oc1ccc(cc1)B(O)O

InChI

1S/C12H21BO3Si/c1-12(2,3)17(4,5)16-11-8-6-10(7-9-11)13(14)15/h6-9,14-15H,1-5H3

InChI key

NVHHEADQQACSCJ-UHFFFAOYSA-N

Application

Reactant involved in:
  • Asymmetric addition reactions with β-substituted cyclic enones
  • Hydroarylation and heterocyclization with phenylpropiolates
  • Double Suzuki-Miyaura coupling reactions

Starting material for the synthesis of red electroluminescent polyfluorenes

Reactant involved in the synthesis of biologically active molecules including:
  • Phenylpyridone derivatives as MCH1R antagonists
  • Atromentin and its O-alkylated derivatives
  • Gelatinases and MT1-MMP inhibitors

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service