Skip to Content
Merck
All Photos(1)

Key Documents

521205

Sigma-Aldrich

4-Ethynyl-1-fluoro-2-methylbenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CH3)C≡CH
CAS Number:
Molecular Weight:
134.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5210 (lit.)

bp

169-170 °C (lit.)

density

1.007 g/mL at 25 °C (lit.)

SMILES string

Cc1cc(ccc1F)C#C

InChI

1S/C9H7F/c1-3-8-4-5-9(10)7(2)6-8/h1,4-6H,2H3

InChI key

GSGKOWLZNQXGAI-UHFFFAOYSA-N

General description

4-Ethynyl-1-fluoro-2-methylbenzene has been reported to afford 1,4-disubstituted 1,2,3-triazole on reaction with azide under typical Cu(I)-catalyzed azide alkyne cycloaddition (CuAAC) conditions.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

100.0 °F - closed cup

Flash Point(C)

37.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janina Moeker et al.
Journal of medicinal chemistry, 57(8), 3522-3531 (2014-04-03)
Carbonic anhydrase IX (CA IX) is a target for hypoxic cancer therapies, and the discovery of CA IX selective ligands is imperative for the development of these agents. Primary sulfonamides are broad specificity inhibitors of CA enzymes, while secondary sulfonamides

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service