Skip to Content
Merck
All Photos(1)

Key Documents

520942

Sigma-Aldrich

2-Bromobenzaldehyde diethyl acetal

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH(OC2H5)2
CAS Number:
Molecular Weight:
259.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.5156 (lit.)

bp

116 °C/0.7 mmHg (lit.)

density

1.285 g/mL at 25 °C (lit.)

SMILES string

CCOC(OCC)c1ccccc1Br

InChI

1S/C11H15BrO2/c1-3-13-11(14-4-2)9-7-5-6-8-10(9)12/h5-8,11H,3-4H2,1-2H3

InChI key

APDGYQVYBWGBSD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.0 °F - closed cup

Flash Point(C)

112.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective synthesis of 2, 3-epoxy alcohols mediated by a remote sulfinyl group.
Ruano JLG, et al.
Tetrahedron, 66(8), 1581-1585 (2010)
New stibines containing acetal and formyl group: Platinum complex and unexpected oxastibol derivative.
Sharma P, et al.
Journal of Organometallic Chemistry, 691(4), 579-584 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service