Recommended Products
Assay
98%
refractive index
n20/D 1.406 (lit.)
bp
117 °C (lit.)
density
0.734 g/mL at 25 °C (lit.)
SMILES string
CCCN(C)CCC
InChI
1S/C7H17N/c1-4-6-8(3)7-5-2/h4-7H2,1-3H3
InChI key
UVBMZKBIZUWTLV-UHFFFAOYSA-N
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
38.0 °F - closed cup
Flash Point(C)
3.33 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Design and evaluation of switchable-hydrophilicity solvents.
Green Chemistry, 16(3), 1187-1197 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service