Skip to Content
Merck
All Photos(1)

Key Documents

516597

Sigma-Aldrich

Rink acid resin

100-200 mesh, extent of labeling: 0.5-1.5 mmol/g loading, 1 % cross-linked with divinylbenzene

Synonym(s):

(4-[(2,4-Dimethoxyphenyl)hydroxymethyl]phenol, polymer-bound, Rink acid-labile (hydroxymethyl)polystyrene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 347.00

CHF 347.00


Estimated to ship on08 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
CHF 347.00

About This Item

MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

CHF 347.00


Estimated to ship on08 May 2025


Request a Bulk Order

Quality Level

crosslinking

1 % cross-linked with divinylbenzene

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

0.5-1.5 mmol/g loading

particle size

100-200 mesh

application(s)

peptide synthesis

functional group

alcohol

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.COc3ccc(C(O)c4ccc(OCc5ccc(C=C)cc5)cc4)c(OC)c3

InChI

1S/C24H24O4.C10H10.C8H8/c1-4-17-5-7-18(8-6-17)16-28-20-11-9-19(10-12-20)24(25)22-14-13-21(26-2)15-23(22)27-3;1-3-9-5-7-10(4-2)8-6-9;1-2-8-6-4-3-5-7-8/h4-15,24-25H,1,16H2,2-3H3;3-8H,1-2H2;2-7H,1H2

InChI key

XFCYBTFZHOFUSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Acid labile resin used in Fmoc-based solid phase peptide synthesis (SPPS).
  • Mild acidic cleavage conditions lead to the release of the peptide acid where fully protected peptides can be released if desired.
  • Cleavage of the final protected peptide can be achieved in as little as 10% AcOH in DCM or 1% TFA in DCM.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service