Skip to Content
Merck
All Photos(1)

Key Documents

516554

Sigma-Aldrich

3-Fluorobenzyl isocyanate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 360.00

CHF 360.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
CHF 360.00

About This Item

Linear Formula:
FC6H4CH2NCO
CAS Number:
Molecular Weight:
151.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 360.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

refractive index

n20/D 1.506 (lit.)

bp

209 °C (lit.)

functional group

amine
fluoro
isocyanate

storage temp.

2-8°C

SMILES string

Fc1cccc(CN=C=O)c1

InChI

1S/C8H6FNO/c9-8-3-1-2-7(4-8)5-10-6-11/h1-4H,5H2

InChI key

PHRJTGPFEAUEBC-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

195.0 °F - closed cup

Flash Point(C)

90.56 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, synthesis and structure-activity relationships of 1, 3, 4-oxadiazole derivatives as novel inhibitors of glycogen synthase kinase-3?.
Saitoh M, et al.
Bioorganic & Medicinal Chemistry, 17(5), 2017-2029 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service