Skip to Content
Merck
All Photos(2)

Documents

511420

Sigma-Aldrich

Poly(3-butylthiophene-2,5-diyl)

regiorandom

Synonym(s):

P3BT

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

color

red

solubility

chloroform, methylene chloride, toluene, and THF: soluble

fluorescence

λex 419 nm; λem 550 nm in chloroform

InChI

1S/C10H16S/c1-4-5-6-10-7-8(2)11-9(10)3/h7H,4-6H2,1-3H3

InChI key

DUOSBQJOYVIVOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Poly(3-butylthiophene-2,5-diyl) (P3BT) is an alkylthiophene based conducting polymer that can be used as a donor molecule in the development of organic electronics. It is a π-conjugating polymer with a π-π stacking distance of 0.395 nm.

Application

P3BT can act as a hole transporting layer (HTL) which can potentially be used in the fabrication of organic field effect transistors (OFETs), chemical sensors, rechargeable batteries and polymeric solar cells (PSCs).

Packaging

Packaged in glass bottles

Citation

For solid state properties see J. Am. Chem. Soc. .

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of alkyl side-chain length and solvent on the luminescent characteristics of poly (3-n-alkylthiophene)
Shin C and Lee H
Synthetic Metals, 140(2-3), 177-181 (2004)
All-organic thin-film transistors patterned by means of selective electropolymerization
Becker E, et al.
Applied Physics Letters, 83(19), 4044-4046 (2003)
High molecular weights, polydispersities, and annealing temperatures in the optimization of bulk-heterojunction photovoltaic cells based on poly (3-hexylthiophene) or poly (3-butylthiophene)
Hiorns RC, et al.
Advances in Functional Materials, 16(17), 2263-2273 (2006)
Nanofiber preparation by whisker method using solvent-soluble conducting polymers
Samitsu S, et al.
Thin Solid Films, 516(9), 2478-2486 (2008)
Journal of the American Chemical Society, 117, 233-233 (1994)

Articles

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

Novel Graphene‑Based Nanostructures Production, Functionalization, and Engineering

Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.

Progress in solution-processed functional materials leads to thin-film optoelectronic devices for industrial and consumer electronics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service