Skip to Content
Merck
All Photos(1)

Documents

50051

Sigma-Aldrich

Glycine benzyl ester p-toluenesulfonate salt

≥99.0% (T)

Synonym(s):

Benzyl glycinate p-toluenesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2COOCH2C6H5 · CH3C6H4SO3H
CAS Number:
Molecular Weight:
337.39
Beilstein:
3600116
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (T)

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

132-134 °C

solubility

methanol: 0.1 g/mL, clear

application(s)

peptide synthesis

SMILES string

Cc1ccc(cc1)S(O)(=O)=O.NCC(=O)OCc2ccccc2

InChI

1S/C9H11NO2.C7H8O3S/c10-6-9(11)12-7-8-4-2-1-3-5-8;1-6-2-4-7(5-3-6)11(8,9)10/h1-5H,6-7,10H2;2-5H,1H3,(H,8,9,10)

InChI key

WJKJXKRHMUXQSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Lin et al.
Molecules (Basel, Switzerland), 25(19) (2020-10-04)
The discovery of IDO1 and HDAC1 dual inhibitors may provide a novel strategy for cancer treatment by taking advantages of both immunotherapeutic and epigenetic drugs. In this paper, saprorthoquinone (1) and 13 of its analogues from Salvia prionitis Hance were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service