Skip to Content
Merck
All Photos(1)

Key Documents

499757

Sigma-Aldrich

Phenoxyacetonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OCH2CN
CAS Number:
Molecular Weight:
133.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.524 (lit.)

bp

235-238 °C (lit.)

density

1.09 g/mL at 25 °C (lit.)

functional group

nitrile
phenoxy

SMILES string

N#CCOc1ccccc1

InChI

1S/C8H7NO/c9-6-7-10-8-4-2-1-3-5-8/h1-5H,7H2

InChI key

VLLSCJFPVSQXDM-UHFFFAOYSA-N

Application

Phenoxyacetonitrile may be used in the synthesis of:
  • 2,4-dihydroxyphenoxyacetophenones[1]
  • methylthio(phenoxy)acetonitrile[2]
  • 2,4-diamino-5-(3,4,5-trimethoxyphenoxy)pyrimidine[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Simple Procedure for the Preparation of (Methyl-and Phenylthio) aryloxyacetonitriles.
Jonczyk A and Muttar EH
Organic preparations and procedures international, 25(6), 690-693 (1993)
Chemistry of modified flavonoids.
Arkhipov VV, et al.
Chemistry of Heterocyclic Compounds, 33(5), 515-519 (1997)
Synthesis of trimethoprim variations. Replacement of methylene by polar groupings.
Stogryn EL.
Journal of Medicinal Chemistry, 15(2), 200-201 (1972)
Reactions of quinoline and 4-chloroquinoline 1-oxides with phenoxyacetonitrile, chloromethylphenylsulfone, and methyl-thiomethyl-p-tolylsulfone.
Hamana M, et al.
Heterocycles, 25, 229-233 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service