Skip to Content
Merck
All Photos(1)

Documents

494232

Sigma-Aldrich

Vacuum pump oil

viscosity 150 cP (25 °C)(lit.)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41100000

manufacturer/tradename

Leybold

refractive index

n20/D 1.4811 (lit.)

viscosity

150 cP(25 °C)(lit.)

bp

389 °C (lit.)

density

0.88 g/mL at 25 °C (lit.)

General description

High quality oil withstands acids, alkalis, and other corrosives better than standard grade vacuum pump oils.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

482.0 °F - Cleveland open cup

Flash Point(C)

250 °C - Cleveland open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service