Recommended Products
isotopic purity
95 atom % 18O
refractive index
n20/D 1.447 (lit.)
bp
210 °C (lit.)
mp
2-3 °C (lit.)
density
1.182 g/mL at 25 °C
mass shift
M+2
SMILES string
[H]C(N)=[18O]
InChI
1S/CH3NO/c2-1-3/h1H,(H2,2,3)/i3+2
InChI key
ZHNUHDYFZUAESO-YZRHJBSPSA-N
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Repr. 1B - STOT RE 2 Oral
Target Organs
Blood
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
309.2 °F - closed cup
Flash Point(C)
154 °C - closed cup
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service