Skip to Content
Merck
All Photos(1)

Key Documents

483478

Sigma-Aldrich

Hexyltrimethoxysilane

Synonym(s):

n -Hexyltrimethoxysilane, Trimethoxyhexylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H22O3Si
CAS Number:
Molecular Weight:
206.35
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

SMILES string

CCCCCC[Si](OC)(OC)OC

InChI

1S/C9H22O3Si/c1-5-6-7-8-9-13(10-2,11-3)12-4/h5-9H2,1-4H3

InChI key

CZWLNMOIEMTDJY-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shihan Xu et al.
Scientific reports, 6, 22350-22350 (2016-03-01)
Herein, we report a facile route to synthesize silane coated upconversion nanoparticles (UCNPs@silane) with an ultrathin layer (the thickness: 1-2 nm), which not only provides good biocompatibility, but also affords hydrophobic interspace to load organic molecules to realize multifunctions. Besides the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service