Skip to Content
Merck
All Photos(2)

Key Documents

480827

Sigma-Aldrich

Tricyclohexyltin chloride

95%

Synonym(s):

Chlorotricyclohexyltin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)3SnCl
CAS Number:
Molecular Weight:
403.62
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

123-125 °C (lit.)

SMILES string

Cl[Sn](C1CCCCC1)(C2CCCCC2)C3CCCCC3

InChI

1S/3C6H11.ClH.Sn/c3*1-2-4-6-5-3-1;;/h3*1H,2-6H2;1H;/q;;;;+1/p-1

InChI key

OJVZYIKTLISAIH-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service