Skip to Content
Merck
All Photos(1)

Key Documents

479039

Sigma-Aldrich

Trichloro(dichloromethyl)silane

96%

Synonym(s):

(Dichloromethyl)trichlorosilane, Trichloro(dichloromethyl)silane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2CHSiCl3
CAS Number:
Molecular Weight:
218.37
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

refractive index

n20/D 1.474 (lit.)

bp

144-146 °C (lit.)

density

1.552 g/mL at 25 °C (lit.)

SMILES string

ClC(Cl)[Si](Cl)(Cl)Cl

InChI

1S/CHCl5Si/c2-1(3)7(4,5)6/h1H

InChI key

LQUJCRPJNSUWKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service