Skip to Content
Merck
All Photos(1)

Documents

478385

Sigma-Aldrich

3-Chloro-4-fluorophenyl isocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)NCO
CAS Number:
Molecular Weight:
171.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.541 (lit.)

bp

44 °C/17 mmHg (lit.)

density

1.377 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(cc1Cl)N=C=O

InChI

1S/C7H3ClFNO/c8-6-3-5(10-4-11)1-2-7(6)9/h1-3H

InChI key

XVIPJBUXMFLHSI-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service