Skip to Content
Merck
All Photos(1)

Documents

477672

Sigma-Aldrich

1,3,5,7,9,11,14-Heptacyclohexyltricyclo[7.3.3.15,11]heptasiloxane-3,7,14-triol

97%

Synonym(s):

Heptacyclohexyl trisilanol POSS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H80O12Si7
CAS Number:
Molecular Weight:
973.68
UNSPSC Code:
12162002
PubChem Substance ID:

Assay

97%

form

solid

mp

109 °C (lit.)

SMILES string

O[Si]1(O[Si]2(O[Si](O)(O[Si]3(O[Si](O)(O[Si](O1)(O[Si](O2)(O3)C4CCCCC4)C5CCCCC5)C6CCCCC6)C7CCCCC7)C8CCCCC8)C9CCCCC9)C%10CCCCC%10

InChI

1S/C42H80O12Si7/c43-55(36-22-8-1-9-23-36)46-58(39-28-14-4-15-29-39)48-56(44,37-24-10-2-11-25-37)50-60(41-32-18-6-19-33-41)51-57(45,38-26-12-3-13-27-38)49-59(47-55,40-30-16-5-17-31-40)53-61(52-58,54-60)42-34-20-7-21-35-42/h36-45H,1-35H2

InChI key

GAUUYVMFTLBDED-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service