Skip to Content
Merck
All Photos(1)

Documents

477400

Sigma-Aldrich

(Acetonitrile)trichlorobis(triphenylphosphine)rhenium(III)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)3P]2Re(CH3CN)Cl3
CAS Number:
Molecular Weight:
858.19
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

97%

reaction suitability

core: rhenium
reagent type: catalyst

mp

210 °C (dec.) (lit.)

SMILES string

CC#N.Cl[Re](Cl)Cl.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6

InChI

1S/2C18H15P.C2H3N.3ClH.Re/c2*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2-3;;;;/h2*1-15H;1H3;3*1H;/q;;;;;;+3/p-3

InChI key

SEHZOVVCPGBXNW-UHFFFAOYSA-K

Application

Catalyst for addition of tetrahalomethanes to alkenes and 1-omega-dienes

Reactant for synthesis of:
  • Re(III) complexes containing 2-benzoylpyridine
  • Rhenium(IV) and (V) complexes
  • Rhenium triphosphorus macrocycle complexes and reactions with alkenes

Reactant for rhenium chemistry of azooximes

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service