Skip to Content
Merck
All Photos(1)

Key Documents

46880

Sigma-Aldrich

Fluorene

≥99.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10
CAS Number:
Molecular Weight:
166.22
Beilstein:
1363491
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥99.0% (HPLC)

bp

298 °C (lit.)

mp

111-114 °C (lit.)
113-115 °C

SMILES string

C1c2ccccc2-c3ccccc13

InChI

1S/C13H10/c1-3-7-12-10(5-1)9-11-6-2-4-8-13(11)12/h1-8H,9H2

InChI key

NIHNNTQXNPWCJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

λmax. 261 nm, log ε 4.28 (heptane)

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

303.8 °F - closed cup

Flash Point(C)

151.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New well-defined poly (2, 7-fluorene) derivatives: photoluminescence and base doping.
Ranger M, et al.
Macromolecules, 30(25), 7686-7691 (1997)
Qu Zhang et al.
Chemical communications (Cambridge, England), 51(50), 10182-10185 (2015-05-28)
Three amino-functionalized fluorene oligomers with different solubility were developed as cathode interfacial materials for inverted polymer solar cells (I-PSCs). By side chain design, we solved the interface layer erosion problem for I-PSCs, and the devices exhibit a power conversion efficiency
Priyanka Chaudhary et al.
Environmental monitoring and assessment, 187(6), 391-391 (2015-06-01)
Polyaromatic hydrocarbons (PAHs) utilizing bacteria were isolated from soils of seven sites of Mathura refinery, India. Twenty-six bacterial strains with different morphotypes were isolated. These strains were acclimatized to utilize a mixture of four polycyclic aromatic hydrocarbons, i.e., anthracene, fluorene
Spectroscopy and photochemistry of fluorene at a silica gel/air interface.
Barbas JT, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 109(3), 229-236 (1997)
Electroluminescent poly (fluorene-co-thiophene-S, S-dioxide): synthesis, characterisation and structure-property relationships.
Pasini M, et al.
Journal of Materials Chemistry, 13(4), 807-813 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service