Skip to Content
Merck
All Photos(1)

Key Documents

468460

Sigma-Aldrich

1,4-Dimethoxynaphthalene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12O2
CAS Number:
Molecular Weight:
188.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

86-90 °C (lit.)

SMILES string

COc1ccc(OC)c2ccccc12

InChI

1S/C12H12O2/c1-13-11-7-8-12(14-2)10-6-4-3-5-9(10)11/h3-8H,1-2H3

InChI key

FWWRTYBQQDXLDD-UHFFFAOYSA-N

Application

1,4-Dimethoxynaphthalene may be used in the preparation of 1,7-dideoxy-3-demethylprekinamycin[1] and 3-acetoxy-2-bromo-1,4-dimethoxynaphthalene.[2]3-acetoxy-2-bromo-1,4-dimethoxynaphthalene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel haloacetoxylation of 1, 4-dimethoxynaphthalenes using hypervalent iodine chemistry.
Evans PA and Brandt TA.
Tetrahedron Letters, 27(36), 6443-6446 (1996)
William Williams et al.
The Journal of organic chemistry, 62(13), 4364-4369 (1997-06-27)
2-(Diazobenzyl)-p-naphthoquinone was synthesized from 2-benzyl-1,4-dimethoxynaphthalene by cerric ammonium nitrate oxidation to 2-benzyl-p-naphthoquinone followed by diazo transfer with tosyl azide. 1,7-Dideoxy-3-demethylprekinamycin was prepared from 1,4-dimethoxynaphthalene by bromination, lithiation, and condensation with acetanthranil to give 2-(2'-N-acetaminobenzoyl)-1,4-dimethoxynaphthalene, which, following deacylation, was subjected to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service