Skip to Content
Merck
All Photos(1)

Documents

467405

Sigma-Aldrich

1-Ethyl-2-iodobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4I
CAS Number:
Molecular Weight:
232.06
MDL number:
UNSPSC Code:
12352103

Assay

97%

refractive index

n20/D 1.595 (lit.)

bp

64 °C/5 mmHg (lit.)

density

1.618 g/mL at 25 °C (lit.)

SMILES string

CCc1ccccc1I


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service