Skip to Content
Merck
All Photos(1)

Key Documents

467251

Sigma-Aldrich

Ethyl 2-isocyanatobenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCNC6H4CO2C2H5
CAS Number:
Molecular Weight:
191.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

28-31 °C (lit.)

SMILES string

CCOC(=O)c1ccccc1N=C=O

InChI

1S/C10H9NO3/c1-2-14-10(13)8-5-3-4-6-9(8)11-7-12/h3-6H,2H2,1H3

InChI key

HXVGHXDQGUPUKY-UHFFFAOYSA-N

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service