Skip to Content
Merck
All Photos(1)

Key Documents

46083

Sigma-Aldrich

Ethylenediaminetetrapropionic acid

≥97.0% (calc. based on dry substance)

Synonym(s):

3,3′,3″,3″′-(Ethylenedinitrilo)tetrapropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24N2O8
CAS Number:
Molecular Weight:
348.35
Beilstein:
1716926
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

≥97.0% (calc. based on dry substance)

form

solid

impurities

5-10% water

mp

178-181 °C (dec.)

SMILES string

OC(=O)CCN(CCN(CCC(O)=O)CCC(O)=O)CCC(O)=O

InChI

1S/C14H24N2O8/c17-11(18)1-5-15(6-2-12(19)20)9-10-16(7-3-13(21)22)8-4-14(23)24/h1-10H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24)

InChI key

KWYJDIUEHHCHCZ-UHFFFAOYSA-N

Other Notes

Complexing agent; the stability constant with Cu(II) (about 18) is much larger than with other cations[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L.H.J. Lajunen et al
Acta Chemica Scandinavica, 43, 2-2 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service