460737
3,6-Bis(2-methyl-2-morpholinopropionyl)-9-octylcarbazole
97%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C36H51N3O4
CAS Number:
Molecular Weight:
589.81
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
Assay
97%
mp
116-120 °C (lit.)
SMILES string
CCCCCCCCn1c2ccc(cc2c3cc(ccc13)C(=O)C(C)(C)N4CCOCC4)C(=O)C(C)(C)N5CCOCC5
Application
Photoinitiator
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service