Skip to Content
Merck
All Photos(1)

Key Documents

458880

Sigma-Aldrich

3-Bromo-2,4,6-trimethylaniline

98%

Synonym(s):

3-Bromo-2,4,6-trimethylbenzenamine

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CHF 270.00

CHF 270.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
CHF 270.00

About This Item

Linear Formula:
BrC6H(CH3)3NH2
CAS Number:
Molecular Weight:
214.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

CHF 270.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

(Powder or Solid or Crystals or Semi-Solid or Liquid)

color

, Light Brown to Dark Brown to Dark Purple

refractive index

n20/D 1.575 (lit.)

bp

172-173 °C/24 mmHg (lit.)

mp

34.5-39.3 °C (lit.)

density

1.313 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Cc1cc(C)c(Br)c(C)c1N

InChI

1S/C9H12BrN/c1-5-4-6(2)9(11)7(3)8(5)10/h4H,11H2,1-3H3

InChI key

MVLMPTBHZPYDBZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdalena Markowicz et al.
Journal of biological physics, 38(4), 637-656 (2012-11-13)
The interactions between dendrimers and different types of drugs are nowadays one of the most actively investigated areas of the pharmaceutical sciences. The interactions between dendrimers and drugs can be divided into: internal encapsulation, external electrostatic interaction, and covalent conjugation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service