Skip to Content
Merck
All Photos(1)

Key Documents

458503

Sigma-Aldrich

Bis[2-[2-(1-methylethyl)-3-oxazolidinyl]ethyl] 1,6-hexanediylbiscarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H46N4O6
CAS Number:
Molecular Weight:
486.65
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

refractive index

n20/D 1.485 (lit.)

viscosity

110 poise(20 °C)(lit.)

bp

200 °C (lit.)

solubility

hydrocarbons, esters and ketones: soluble

density

1.085 g/mL at 25 °C (lit.)

SMILES string

CC(C)C1OCCN1CCOC(=O)NCCCCCCNC(=O)OCCN2CCOC2C(C)C

General description

Forms hydroxyl and amino groups upon hydrolysis.

Application

Moisture scavenger, polymer additive.
Latent cross-linking agent for polyurethane prepolymers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service