Skip to Content
Merck
All Photos(1)

Key Documents

458465

Sigma-Aldrich

N-(tert-Butoxycarbonyl)-L-valine N′-methoxy-N′-methylamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH[NHCO2C(CH3)3]CON(OCH3)CH3
CAS Number:
Molecular Weight:
260.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

optical activity

[α]23/D −18.5°, c = 1 in methanol

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.456 (lit.)

bp

248 °C (lit.)

density

1.029 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

SMILES string

CON(C)C(=O)[C@@H](NC(=O)OC(C)(C)C)C(C)C

InChI

1S/C12H24N2O4/c1-8(2)9(10(15)14(6)17-7)13-11(16)18-12(3,4)5/h8-9H,1-7H3,(H,13,16)/t9-/m0/s1

InChI key

RRBFCGUIFHFYQK-VIFPVBQESA-N

Application

α-Amino Weinreb amide.[1][2][3][4][5][6][7][8]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kosynkina, L. et al.
Tetrahedron Letters, 35, 5173-5173 (1994)
Saari, W.S. Fischer, T.E.
Synthesis, 453-453 (1990)
Angelastro, M.R. at al.
Tetrahedron Letters, 33, 3265-3265 (1992)
Kim, B.M. et al.
Tetrahedron Letters, 35, 5153-5153 (1994)
Lucet, D. et al.
Tetrahedron Asymmetry, 7, 985-985 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service