Skip to Content
Merck
All Photos(2)

Key Documents

457183

Sigma-Aldrich

Ammonium hexafluorostannate

≥99.99% trace metals basis

Synonym(s):

Diammonium hexafluorostannate, Diammonium tin hexafluoride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
CHF 94.50

CHF 94.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
CHF 94.50

About This Item

Linear Formula:
(NH4)2SnF6
CAS Number:
Molecular Weight:
266.76
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
crystalline

CHF 94.50


Check Cart for Availability

Request a Bulk Order

grade

for analytical purposes

Assay

≥99.99% trace metals basis

form

crystalline

reaction suitability

reagent type: catalyst
core: tin

impurities

≤100.0 ppm Trace Metal Analysis

SMILES string

[NH4+].[NH4+].F[Sn--](F)(F)(F)(F)F

InChI

1S/6FH.2H3N.Sn/h6*1H;2*1H3;/q;;;;;;;;+4/p-4

InChI key

UWWCEQYTGCAXHS-UHFFFAOYSA-J

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zeng Chen et al.
Nanoscale research letters, 11(1), 295-295 (2016-06-15)
SnO2 nanosheet-structured films were prepared on a fluorine-doped tin oxide (FTO) substrate using ZnO nanosheet as template. The as-prepared SnO2 nanosheets contained plenty of nano-voids and were generally vertical to the substrate. TiO2 nanoparticles were homogeneously deposited into the intervals

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service