Skip to Content
Merck
All Photos(1)

Key Documents

452467

Sigma-Aldrich

3,4-Dimethoxybenzenesulfonyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3SO2Cl
CAS Number:
Molecular Weight:
236.67
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

67-70 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1ccc(cc1OC)S(Cl)(=O)=O

InChI

1S/C8H9ClO4S/c1-12-7-4-3-6(14(9,10)11)5-8(7)13-2/h3-5H,1-2H3

InChI key

RSJSYCZYQNJQPY-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Limitations of the Transition State Variation Model. Part 8. Dual Reaction Channels for Solvolyses of 3, 4-Dimethoxybenzenesulfonyl Chloride
Koo IS, et al.
Bull. Korean Chem. Soc., 28(12), 2377-2377 (2007)
A M Gilbert et al.
Journal of medicinal chemistry, 43(6), 1223-1233 (2000-03-29)
A series of xanthine sulfonamides is presented as a class of calcitonin (CT) inducers - a potentially new method for treating diseases associated with postmenopausal bone loss such as osteoporosis. We have found that certain di-n-butylxanthine sulfonamides 4 upregulate CT
K Nicolaou et al.
Chemistry & biology, 7(12), 979-992 (2001-01-04)
NADH:ubiquinone oxidoreductase (complex I) is the first of three large enzyme complexes located in the cell's inner mitochondrial membrane which form the electron transport chain that carries electrons from NADH to molecular oxygen during oxidative phosphorylation. There is significant interest
Monomethylthio analogs of 1-(2, 4, 5-trimethoxyphenyl)-2-aminopropane.
Jacob III P, et al.
Journal of Medicinal Chemistry, 30(10), 1235-1239 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service