447994
Flurbiprofen
97%
Synonym(s):
(±)-2-Fluoro-α-methyl-4-biphenylacetic acid, L-790,330
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
97%
mp
110-112 °C (lit.)
SMILES string
CC(C(O)=O)c1ccc(c(F)c1)-c2ccccc2
InChI
1S/C15H13FO2/c1-10(15(17)18)12-7-8-13(14(16)9-12)11-5-3-2-4-6-11/h2-10H,1H3,(H,17,18)
InChI key
SYTBZMRGLBWNTM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Fluibiprofen is a cyclooxygenase (COX) inhibitor, which is an enzyme responsible for the conversion of arachidonic acid to prostaglandin G2 (PGG2) and PGG2 to prostaglandin H2 (PGH2) in the prostaglandin synthesis pathway. This decreases the prostaglandins which cause inflammation, pain, swelling and fever. Flurbiprofen inhibits the activity of both COX-1 and -2. The S enantiomer inhibits prostaglandin synthesis and has both anti-inflammatory and analgesic activity, while the R enantiomer does not inhibit prostaglandin synthesis and displays only analgesic activity.
Application
Product No.
Description
Pricing
replaced by
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
European journal of medicinal chemistry, 43(12), 2688-2698 (2008-04-09)
A series of 1,3,4-oxadiazole/thiadiazole and 1,2,4-triazole derivatives of biphenyl-4-yloxy acetic acid were synthesized in order to obtain new compounds with potential anti-inflammatory activity, analgesic activity and lower ulcerogenic potential. All compounds were evaluated for their anti-inflammatory activity by the carrageenan
Bioorganic & medicinal chemistry letters, 18(3), 918-922 (2008-01-10)
A series of 3-(4-biphenyl)-5-substituted phenyl-2-pyrazolines (2a-h) and 1-benzoyl-3-(4-biphenyl)-5-substituted phenyl-2-pyrazolines (3a-h) were synthesized by condensation of chalcones with hydrazine hydrate in solvent system ethanol and DMF. The newly synthesized compounds were screened for their anti-inflammatory and analgesic activity, and were compared
Bioorganic & medicinal chemistry, 18(6), 2204-2218 (2010-03-02)
Chinese herbs were screened for compounds which may be active against four targets involved in inflammation, using pharmacophore-assisted docking. Multiple LigandScout (LS) pharmacophores built from ligand-receptor complexes in the protein databank (PDB) were first employed to select compounds. These compounds
Journal of medicinal chemistry, 48(18), 5705-5720 (2005-09-02)
Flurbiprofen, a nonsteroidal antiinflammatory drug (NSAID), has been recently described to selectively inhibit beta-amyloid(1)(-)(42) (Abeta42) secretion, the most toxic component of the senile plaques present in the brain of Alzheimer patients. The use of this NSAID in Alzheimer's disease (AD)
European journal of medicinal chemistry, 43(10), 2056-2066 (2007-11-21)
Several 3,6-disubstituted-1,2,4-triazolo-[3,4-b]-1,3,4-thiadiazoles were prepared by condensation of 4-amino-5-substituted-3-mercapto-(4H)-1,2,4-triazoles (3a,b) with various substituted aromatic acids and aryl/alkyl isothiocyanates through a one-pot reaction. These compounds were investigated for their anti-inflammatory, analgesic, ulcerogenic, lipid peroxidation, antibacterial and antifungal activities. Some of the synthesized
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service