Skip to Content
Merck
All Photos(1)

Key Documents

438944

Sigma-Aldrich

1-(2-Naphthoyl)imidazole

95%

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H10N2O
CAS Number:
Molecular Weight:
222.24
Beilstein:
5431262
MDL number:
UNSPSC Code:
12352200

Assay

95%

mp

74-78 °C (lit.)

SMILES string

O=C(c1ccc2ccccc2c1)n3ccnc3

InChI

1S/C14H10N2O/c17-14(16-8-7-15-10-16)13-6-5-11-3-1-2-4-12(11)9-13/h1-10H

InChI key

FEYNPDBWOXBMOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service