Skip to Content
Merck
All Photos(1)

Key Documents

434698

Sigma-Aldrich

Poly(ethylene-co-1-butene)

melt index 0.8 g/10 min (190°C/2.16kg)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2)x[CH2CH(C2H5)]y
CAS Number:
MDL number:
UNSPSC Code:
12162002

melt index

0.8 g/10 min (190°C/2.16kg)

contains

antioxidant (varying amounts)

hardness

30 (Shore D, ASTM 2240)

mp

60 °C

transition temp

softening point 72 °C (Vicat, 200g, ASTM D 1525)

density

0.88 g/mL at 25 °C

Mw/Mn

~2.0

elongation

ASTM D 412 - 620% (at break)

SMILES string

C=C.CCC=C

Looking for similar products? Visit Product Comparison Guide

Application

Additive for polyethylene and polypropylene used in injection, extrusion and blow molding.

Features and Benefits

Linear copolymer.
Exhibits plastic and elastic properties. Low temperature impact modifier, high clarity, reduces stress cracking and has low temperature heat sealability. Narrow molecular weight and composition distributions.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service