Skip to Content
Merck
All Photos(1)

Documents

43158

Sigma-Aldrich

(R)-(−)-2-[2-(Diphenylphosphino)phenyl]-4-phenyl-2-oxazoline

≥97.0% (31P-NMR)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H22NOP
CAS Number:
Molecular Weight:
407.44
Beilstein:
7547072
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥97.0% (31P-NMR)

form

solid

optical activity

[α]20/D −29.5±3°, c = 1% in chloroform

SMILES string

C1OC(=N[C@@H]1c2ccccc2)c3ccccc3P(c4ccccc4)c5ccccc5

InChI

1S/C27H22NOP/c1-4-12-21(13-5-1)25-20-29-27(28-25)24-18-10-11-19-26(24)30(22-14-6-2-7-15-22)23-16-8-3-9-17-23/h1-19,25H,20H2/t25-/m0/s1

InChI key

OKUTYUNUKKPYJS-VWLOTQADSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Chiral ligand employed in enantioselective Pd-catalyzed allylic substitution reactions

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.M.J. Williams et al.
Tetrahedron Letters, 34, 3149-3149 (1993)
P. von Matt et al.
Angewandte Chemie (International Edition in English), 105, 614-614 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service