Skip to Content
Merck
All Photos(3)

Key Documents

423122

Sigma-Aldrich

4-tert-Butylphthalonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3-1,2-(CN)2
CAS Number:
Molecular Weight:
184.24
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

98%

form

powder, crystals or chunks

bp

165-168 °C/6 mmHg (lit.)

mp

49-51 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC(C)(C)c1ccc(C#N)c(c1)C#N

InChI

1S/C12H12N2/c1-12(2,3)11-5-4-9(7-13)10(6-11)8-14/h4-6H,1-3H3

InChI key

LOTMIRVNJTVTSU-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Merve Pamukçu Polat et al.
Dalton transactions (Cambridge, England : 2003), 48(25), 9194-9204 (2019-06-04)
Unsymmetrically substituted phthalocyanines were synthesized by the well-known statistical condensation method using two differently substituted precursors, 4-tert-butylphthalonitrile and 4-(4-pyrrol-1-yl)phenoxyphthalonitrile. Spin-coated thin films of these compounds were subsequently exposed to varying concentrations of methanol, ethanol and butanol between 25 and 150
Gauta Gold Matlou et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 214, 49-57 (2019-02-15)
In this study, a novel asymmetric cinnamic acid zinc phthalocyanine (ZnPc, 1) containing three tert-butyl substituents is reported. The asymmetric ZnPc (1) is further linked to amino functionalized magnetic nanoparticles (AMNPs) (1-AMNPs) and to cysteine functionalized silver nanoparticles (cys-AgNPs) (1-cys-AgNPs)
Min Wang et al.
Nature communications, 10(1), 3602-3602 (2019-08-11)
Molecular catalysts that combine high product selectivity and high current density for CO2 electrochemical reduction to CO or other chemical feedstocks are urgently needed. While earth-abundant metal-based molecular electrocatalysts with high selectivity for CO2 to CO conversion are known, they

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service