Skip to Content
Merck
All Photos(1)

Key Documents

422614

Sigma-Aldrich

(R)-(+)-2-Pyrrolidone-5-carboxylic acid

95%

Synonym(s):

D-5-Oxo-2-pyrrolidinecarboxylic acid, D-Pyroglutamic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO3
CAS Number:
Molecular Weight:
129.11
Beilstein:
82133
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

optical activity

[α]22/D +10°, c = 1.5 in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

155-156 °C (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)[C@H]1CCC(=O)N1

InChI

1S/C5H7NO3/c7-4-2-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m1/s1

InChI key

ODHCTXKNWHHXJC-GSVOUGTGSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica Fiori et al.
Journal of chromatography. A, 1562, 115-122 (2018-06-04)
Monomeric catechins are important compounds in green tea accounting for potential bioactivity against a wide range of diseases. Besides catechins, l-Theanine (γ-glutamylethylamide), a characteristic amino acid in tea leaves, has become a further focus of the phytochemical research for the
Daniel Stettin et al.
Metabolites, 10(4) (2020-04-09)
The development of improved mass spectrometers and supporting computational tools is expected to enable the rapid annotation of whole metabolomes. Essential for the progress is the identification of strengths and weaknesses of novel instrumentation in direct comparison to previous instruments.
Makoto Ariyoshi et al.
Scientific reports, 7, 43911-43911 (2017-03-08)
D-Amino acids are enantiomers of L-amino acids and have recently been recognized as biomarkers and bioactive substances in mammals, including humans. In the present study, we investigated functions of the novel mammalian mitochondrial protein 9030617O03Rik and showed decreased expression under

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service