Skip to Content
Merck
All Photos(1)

Key Documents

417351

Sigma-Aldrich

4-Hydroxy-6-methylcoumarin

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O3
CAS Number:
Molecular Weight:
176.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

261-264 °C (lit.)

functional group

ester
ketone

SMILES string

Cc1ccc2OC(=O)C=C(O)c2c1

InChI

1S/C10H8O3/c1-6-2-3-9-7(4-6)8(11)5-10(12)13-9/h2-5,11H,1H3

InChI key

WIRGBZBGYNIZIB-UHFFFAOYSA-N

Application

4-Hydroxy-6-methylcoumarin may be used for the in situ generation of α-methylenechromanes, α-methylenequinoline and ortho-quinone methides, via Knoevenagel reaction.[1] It may be used for the preparation of 8-methyl-2-phenyl-4H-furo-[3,2-c]benzopyran-4-one and 6-methyl-2-phenyl-4H-furo-[2,3-b]benzopyran-4-one.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Efficient One-Step Synthesis of 2-AryI furans by Ceric Ammonium Nitrate (CAN)-Mediated Cycloaddition of 1, 3-Dicarbonyl Compounds to Alkynes.
Lee YR, et al.
Bull. Korean Chem. Soc., 19(10), 1081-1081 (1998)
José C J M D S Menezes et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 20(42), 13644-13655 (2014-08-30)
In view of increasing demands for efficient photosensitizers for photodynamic therapy (PDT), we herein report the synthesis and photophysical characterizations of new chlorin e6 trimethyl ester and protoporphyrin IX dimethyl ester dyads as free bases and Zn(II) complexes. The synthesis of these
Jin Kyu Kang et al.
Molecules (Basel, Switzerland), 25(19) (2020-10-01)
Coumarins are natural products with promising pharmacological activities owing to their anti-inflammatory, antioxidant, antiviral, anti-diabetic, and antimicrobial effects. Coumarins are present in many plants and microorganisms and have been widely used as complementary and alternative medicines. To date, the pharmacological
Mahla Mirali et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 969, 149-161 (2014-09-01)
A systematic set of optimization experiments was conducted to design an efficient extraction and analysis protocol for screening six different sub-classes of phenolic compounds in the seed coat of various lentil (Lens culinaris Medik.) genotypes. Different compounds from anthocyanidins, flavan-3-ols

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service