Skip to Content
Merck
All Photos(1)

Documents

417122

Sigma-Aldrich

N,N-Diethylnipecotamide

98%

Synonym(s):

N,N-Diethyl-3-piperidinecarboxamide, Piperidine-3-carboxylic acid diethylamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O
CAS Number:
Molecular Weight:
184.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

liquid

refractive index

n20/D 1.489 (lit.)

bp

111 °C/6.1 mmHg (lit.)

density

0.989 g/mL at 25 °C (lit.)

SMILES string

CCN(CC)C(=O)C1CCCNC1

InChI

1S/C10H20N2O/c1-3-12(4-2)10(13)9-6-5-7-11-8-9/h9,11H,3-8H2,1-2H3

InChI key

ZXQKYQVJDRTTLZ-UHFFFAOYSA-N

General description

N,N-Diethylnipecotamide is a six-membered bidentate chelating ligand. It participates as ligand in the synthesis of dimeric halocopper(I) complexes.

Application

N,N-Diethylnipecotamide may be used for the preparation of 1-substituted N,N-diethylnipecotamides.
Reactant for synthesis of:
Antituberculosis drugs
Spiroimidazolidinone NPC1L1 inhibitors
Acyl-CoA:cholesterol acyltransferase inhibitors
Alpha 7 nicotinic acetylcholine receptor agonist
Antimycobacterials
Isocyanoamides useful as starting materials in IMCR

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Kovacic et al.
European journal of pharmacology, 47(1), 37-44 (1978-01-01)
Adult male Holtzman rats were trained to barpress on a schedule whereby every fourth press earned a reward of 0.01 ml of sugar-sweetened milk (FR4). After an i.p. injection of LSD (0.1 mg/kg) or DMT (3.2 or 10 mg/kg) such
Stoichiometry, properties and kinetics of aprotic reduction of dioxygen by [LCuX]2 (L= N, N-diethylnipecotamide; X= Cl, Br).
El-Sayed MA.
Inorgorganica Chimica Acta, 197(2), 197-201 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service