Skip to Content
Merck
All Photos(1)

Key Documents

41609

Sigma-Aldrich

(R)-(4-Fluorophenyl)oxirane

technical, ≥90.0% (GC)

Synonym(s):

(R)-4-Fluorostyrene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7FO
CAS Number:
Molecular Weight:
138.14
Beilstein:
4350328
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90.0% (GC)

optical purity

enantiomeric ratio: ≥99.5:0.5

SMILES string

Fc1ccc(cc1)[C@@H]2CO2

InChI

1S/C8H7FO/c9-7-3-1-6(2-4-7)8-5-10-8/h1-4,8H,5H2/t8-/m0/s1

InChI key

ICVNPQMUUHPPOK-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

72.0 °F - closed cup

Flash Point(C)

22.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service